Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST612

Trace elements in glass

NIST® SRM® 612

Sign Into View Organizational & Contract Pricing

Select a Size

4 UNITS
$1,250.00

$1,250.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 UNITS
$1,250.00

About This Item

UNSPSC Code:
41116107

$1,250.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 4 wafers

manufacturer/tradename

NIST®

application(s)

glass & ceramic

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 612 comprises of four wafers, sliced to 3 mm thickness from a hand-pulled rod. The wafers are of oval to circular cross-section with nominal diameter of 12 mm to 14 mm and an approximate mass of 1.4 g. The nominal mass fractions of 61 elements added to the glass matrix are in the range of 10 mg/kg to 80 mg/kg. For more information, please refer to the COA and SDS.
SRM 612_cert

SRM 612_SDS

Application

The Standard Reference Material (SRM) is primarily intended for facilitating the development of chemical methods for the analysis of trace elements in glass.

Features and Benefits

  • NIST certified values for elements are provided and values are traceable to international system of units (SI) and expressed as mass fraction (mg/kg)
  • Non-certified values, additional information, maintenance of certified values along with instructions for use and storage are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Antimony (Sb), Arsenic (As), Barium (Ba), Cadmium (Cd), Chromium (Cr), Iron (Fe), Lead (Pb), Manganese (Mn), Nickel (Ni), Rubidium (Ru), Selenium (Se), Silver (Ag), Strontium (Sr), Thorium (Th), Uranium (U)

Reference/Informational value is provided for the following analytes.
Cobalt (Co), Copper (Cu), Thallium (Th), Titanium (Ti), Strontium (87Sr/86Sr), Boron (B), Cerium (Ce), Dysprosium (Dy), Erbium (Er), Europium (Eu), Gadolinium (Gd), Gold (Au), Lanthanum (La), Lithium (Li), Neodymium (Nd), Potassium (K), Samarium (Sm), Ytterbium (Yb), Uranium (235U/UTotal)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service