Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST459

Beryllium-copper alloy

(17200), NIST SRM 459, (17200)

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$1,210.00

$1,210.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$1,210.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,210.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in the form of chips sized between 0.50 and 1.18 mm sieve openings (35 and 16 mesh). It is intended primarily for use in chemical methods and analysis. For more information, please refer to the SDS and COA.

SRM 459_cert

SRM 459_SDS

Other Notes

Certified for the analytes listed below.
Beryllium (Be), Cobalt (Co), Nickel (Ni), Iron (Fe), Silicon (Si), Aluminium (Al), Tin (Sn), Zinc (Zn), Chromium (Cr), Lead (Pb), Magnesium (Mg)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Manganese (Mn), Silver (Ag), Sulfur (S), Titanium (Ti), Zirconium (Zr), Copper (Cu)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service