Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST3451

Low temperature Seebeck coefficient standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of bar

manufacturer/tradename

NIST®

General description

Each unit of SRM 3451 contains a bar-shaped artifact of chemical formula Bi2Te3+x, measuring approximately 3.5 mm × 2.5 mm × 8.0 mm. The material is non-stoichiometric bismuth telluride, with n-type conductivity and a Bi/Te mole ratio of about2/3.

SRM 3451_cert

SRM 3451_SDS

Application

This SRM is intended to calibrate Seebeck coefficient measurement instruments for bulk materials within the temperature range of 10 K to 390 K.

Features and Benefits

  • Accompanied with certified values, obtained using a Quantum designphysical property measurement system (PPMS) through a differential steady-state(DC) technique; an uncertainty analysis was performed following the guidelines outlined in the ISO Guide.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service