Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2745

Carbon dioxide in nitrogen

NIST SRM 2745, nominal 16% mol/mol

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$13,180.00

$13,180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$13,180.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$13,180.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

packaging

pkg of cylinder

manufacturer/tradename

NIST®

application(s)

air monitoring
environmental

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a primary gas mixture for which the amount-of-substance fraction, expressed as concentration, may be related to secondary working standards. This SRM is intended for the calibration of instruments used for carbon dioxide determinations and for other applications. For more information, please refer to the SDS and COA.

SRM 2745_cert

SRM 2745_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon Dioxide (CO2)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

related product

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service