Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST2573

Lead paint film (red), nominal 1.0 mg/cm2

NIST® SRM® 2573

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 1 +blank

manufacturer/tradename

NIST®

application(s)

coatings

format

matrix material

General description

This Standard Reference Material (SRM) is intended for validation of results from portable, hand-held, X-ray fluorescence analyzers, when testing for lead in paint coatings on interior and exterior building surfaces. A unit of SRM 2573 consists of a white polyester sheet, approximately 7.6 cm wide, 10.2 cm long, and 0.2 mm thick, coated with a single, red-colored paint layer, approximately 0.04 mm thick. Included is one unit of SRM 2570, which is coated with a lead-free, lacquer layer of the same thickness as a lead paint layer and is intended as a blank. All sheets are over-coated with a clear, thin, plastic laminate to protect the surface from abrasion.For more information, please refer to the COA and SDS.

SRM 2573_cert

SRM 2573_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service