Skip to Content
MilliporeSigma
All Photos(2)

Documents

NIST2186I

Potassium phosphate monobasic

NIST® SRM® 2186I

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

NIST standard reference material

packaging

pkg of 30 g

manufacturer/tradename

NIST®

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

application(s)

general analytical

format

neat

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

SRM 2186I is certified as a pH standard for pD(S) determination.

SRM 2186I _SDS

SRM 2186I _CoA

Application

SRM 2186I solution is recommended for the calibration of pH meters to use for the measurement of pD in deuterium oxide.

Other Notes

Certified for the analytes listed below.
Potassium Dihydrogen Phosphate

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service