Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2160

Low alloy steel

NIST® SRM® 2160, carbon & sulfur only

Sign Into View Organizational & Contract Pricing

Select a Size

200 G
$1,440.00

$1,440.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 G
$1,440.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,440.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 200 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in the form of pins that are 4 mm in diameter, 12 mm long and weigh approximately 1 g each. SRM 2159 is primarily intended for use in calibrating instruments used in the determination of carbon and sulfur in steel. For more information, please refer to the COA and SDS.

SRM 2160_cert

SRM 2160_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C), Sulfur (S)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service