Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2137

Boron implant in silicon standard for calibration of concentration in a depth profile

NIST® SRM® 2137

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

packaging

pkg of each

manufacturer/tradename

NIST®

application(s)

semiconductor

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in calibrating the secondary ion response to minor and trace levels of boron in a silicon matrix by the analytical technique of secondary ion mass spectrometry (SIMS). A unit of SRM 2137 consists of a single crystal silicon substrate with a surface rendered disordered by silicon ion implantation. For more information, please refer to the SDS and COA.

SRM 2137_cert

SRM 2137_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Boron (10B)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service