Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST2061

TiAl(NbW) alloy for microanalysis

NIST® SRM® 2061

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of cube

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 2061 contains a single cube ofthe alloy, approximately measuring 2 mm × 2 mm × 2 mm.

SRM 2061_cert

SRM 2061_SDS

Application

TiAl(NbW)alloy standard reference material (SRM) is intended for micro analysis of titanium and aluminum, particularly for aerospace materials and components with similar composition.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction value of titanium, aluminum, niobium, and tungsten. These values are measured through wavelength dispersive X-ray fluorescence spectrometry(WD-XRF) and inductively coupled plasma optical emission spectrometry(ICP-OES).
  • Information values for oxygen andnitrogen are reported as mass fractions.
  • The NIST certificate is provided with expiration certificate, storage, handling,and use instructions.

Other Notes

Certified for the analytes listed below.
Titanium (Ti), Aluminium (Al), Niobium (Nb), Tungsten (W)

Reference/Informational value is provided for the following analytes.
Oxygen (O), Nitrogen (N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service