Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST191D

pH Standards

NIST® SRM® 191d, set of 191d-I + 191d-II

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,420.00

$2,420.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$2,420.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,420.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

packaging

pkg of 1 bottle x 25 g; 1 bottle x 30 g

manufacturer/tradename

NIST®

application(s)

general analytical

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended for use in preparing solutions for calibrating electrodes for pH measuring systems. SRM 191d consists of two components, each prepared to ensure high purity and uniformity:Sodium Bicarbonate, NaHCO3 (191d-I); and Sodium Carbonate, Na2CO3 (191d-II). However, neither SRM component is certified for purity of substance. A unit of SRM 191d consists of 25 g of sodium bicarbonate (191d-I) and 30 g of sodium carbonate (191d-II), each contained in its respective clear glass bottle. For more information, please refer to the COA and SDS.

SRM 191D_cert

SRM 191D_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service