Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1718

Nitrous oxide in air

NIST SRM 1718, nominal 1 μmol/mol

Sign Into View Organizational & Contract Pricing

Select a Size

NIST1718
$8,740.00

$8,740.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
NIST1718
$8,740.00

About This Item

UNSPSC Code:
41116107

$8,740.00


Please contact Customer Service for Availability

Request a Bulk Order

form

gas (compressed)

packaging

pkg of cylinder

General description

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service