Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MGVS100

Medical Viscosity Standard

Medical Viscosity Standard

viscosity 10.00 mPa.s (25.00 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$93.40
500 ML
$400.00

$93.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$93.40
500 ML
$400.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$93.40


Please contact Customer Service for Availability

Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

10.00 mPa.s(25.00 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Find detailed technical data for all Paragon Medical Viscosity Standards on the Paragon Scientific webpage
These products contain antibacterial agent.
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 under UKAS accreditation

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hi. I wonder if it possible to get the viscosity value of MGVS100 at 20C? It would be really helpful.

    1 answer
    1. This line of Medical Viscosity Standards, manufactured by Paragon Scientific Ltd, is measured only at 25 °C and 37 °C.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service