Recommended Products
grade
analytical standard
Quality Level
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
storage temp.
2-8°C
SMILES string
COc1ccc2[nH]cc(CCO)c2c1
InChI
1S/C11H13NO2/c1-14-9-2-3-11-10(6-9)8(4-5-13)7-12-11/h2-3,6-7,12-13H,4-5H2,1H3
InChI key
QLWKTGDEPLRFAT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
5-Methoxytryptophol may be used as an analytical reference standard for the quantification of the analyte in biological samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of pineal research, 27(4), 230-236 (1999-11-07)
The aim of the present study was to determine whether the genetic differences previously reported in ewe plasma melatonin concentrations were correlated with differences in the synthesis and release of other 5-methoxyindoles. To determine if 5-methoxytryptophol (5-ML), which is known
Journal of pineal research, 32(4), 214-218 (2002-05-02)
The duck pineal gland rhythmically produces two 5-methoxyindole compounds, i.e. 5-methoxytryptophol and melatonin. 5-Methoxytryptophol levels are low at night and high during the day, while melatonin concentrations are high at night and low during the day. The melatonin rhythm reflects
Biochemistry and cell biology = Biochimie et biologie cellulaire, 78(4), 447-453 (2000-09-30)
Male Sprague-Dawley rats were randomly divided into four groups. Two of the groups received a single intraperitoneal injection of melatonin and 5-methoxytryptamine (5 mg/kg body weight), respectively, at 9 PM. One group received an intraperitoneal injection of 5-methoxytryptophol (5 mg/kg
General and comparative endocrinology, 131(2), 168-175 (2003-04-08)
Daily patterns of melatonin and 5-methoxytryptophol (5-ML) concentrations and of aryl alkylamine N-acetyltransferase (AA-NAT) and hydroxyindole-O-methyltransferase (HIOMT) activities have been measured in the cerebroid ganglions, visceral ganglions, and ocular tentacles of the gastropod mollusc Helix aspersa maxima. Melatonin concentrations are
Journal of neural transmission (Vienna, Austria : 1996), 108(4), 397-405 (2001-07-28)
Male C57 mice received 10 consecutive daily intraperitoneal injections of melatonin, 5-methoxytryptamine or 5-methoxytryptophol (5mg/kg body weight). Control mice received the alcoholic saline vehicle. All mice were sacrificed 24 hours after the last injection. Following extraction of RNA from peritoneal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service