Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LTR-CV2-RBD-01

SARS-Cov-2 Spike, Receptor Binding Domain (aa 319- aa 541)

pkg of 1 μg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.41

grade

analytical standard

Assay

>95%

mol wt

Mw 26 kDa

packaging

pkg of 1 μg

manufacturer/tradename

InBio

format

neat

storage temp.

−20°C

General description

Source: E. coli
Formulation: Preservative and carrier-free in 20mM Tris, 0.3M NaCl, 6M Urea, pH 8.0. Filtered through 0.22μm filter.
Concentration: xx based on OD 280nm. Extinction Coefficient = 1.286.
Purification: Purified from E. coli by multi-step chromatography.

Notes:
(1) SARS-CoV-2 NCR has a C-terminal 7xHis-tag.
(2) A LoTox product, Endotoxin < 0.03 EU/μg.

Disclaimer

For research and commercial use in vitro: not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service