Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L1275

Sigma-Aldrich

Leuco Patent Blue Violet

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H32N2O6S2Na2
CAS Number:
Molecular Weight:
590.66
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

~98%

SMILES string

[Na].CCN(CC)c1ccc(cc1)C(c2ccc(cc2)N(CC)CC)c3ccc(cc3S(O)(=O)=O)S(O)(=O)=O

InChI

1S/C27H34N2O6S2.Na.H/c1-5-28(6-2)22-13-9-20(10-14-22)27(21-11-15-23(16-12-21)29(7-3)8-4)25-18-17-24(36(30,31)32)19-26(25)37(33,34)35;;/h9-19,27H,5-8H2,1-4H3,(H,30,31,32)(H,33,34,35);;

InChI key

OBZCRBOHRQBSLN-UHFFFAOYSA-N

Application

Useful in enzyme-linked spectrophotometric determinations of hydrogen peroxide.

Other Notes

Reduced form of Patent Blue Violet

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clapp, P.A., and Evans, D.F.
Analytica Chimica Acta, 243, 217-217 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service