Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM621

Cadmium-111Cd solution

IRMM®, certified reference material, 1 M in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

isotopic purity

 111Cd

manufacturer/tradename

JRC

concentration

1 M in nitric acid

application(s)

general analytical

format

single component solution

storage temp.

10-30°C

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM621

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service