Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I0360000

Ipratropium bromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ipratropium bromide monohydrate, Atropine isopropyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$205.00

About This Item

Empirical Formula (Hill Notation):
C20H30BrNO3 · H2O
CAS Number:
Molecular Weight:
430.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

ipratropium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.[Br-].CC(C)[N@+]1(C)[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C20H30NO3.BrH.H2O/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;;/h4-8,14,16-19,22H,9-13H2,1-3H3;1H;1H2/q+1;;/p-1/t16-,17+,18+,19?,21?;;

InChI key

KEWHKYJURDBRMN-XSAPEOHZSA-M

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ipratropium bromide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maciej Ciebiada et al.
Allergy and asthma proceedings, 35(5), 72-79 (2014-10-09)
Persistent airways obstruction (PAO) may affect some patients with severe asthma and may significantly worsen the prognosis. This study was designed to detect risk factors associated with persistent airflow limitation in nonsmoking adult patients with severe asthma. A total of
Sharath S Hegde et al.
The Journal of pharmacology and experimental therapeutics, 351(1), 190-199 (2014-08-08)
The objective of the present studies was to characterize the pharmacologic properties of GSK-961081 [TD-5959; (R)-1-(3-((2-chloro-4-(((2-hydroxy-2-(8-hydroxy-2-oxo-1,2-dihydroquinolin-5-yl)ethyl)amino)methyl)-5-methoxyphenyl)amino)-3-oxopropyl) piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamate], a novel first-in-class inhaled bifunctional compound possessing both muscarinic antagonist (MA) and β2-adrenoceptor agonist (BA) properties (MABA). In competition radioligand binding studies
Moritz Beck-Broichsitter et al.
Journal of pharmaceutical sciences, 103(8), 2585-2589 (2014-07-06)
Nebulization of active pharmaceutical ingredient (API) solutions is a well-established means to achieve pulmonary drug deposition. The current study identified the impact of formulation variables on the aerosolization performance of the eFlow(®) rapid with special respect to optimized lung application.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service