Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F4166

Sigma-Aldrich

Sodium formate

BioXtra

Synonym(s):

Formic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein:
3595134
EC Number:
MDL number:
UNSPSC Code:
12352103

product line

BioXtra

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

259-262 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.001%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey Zakharov et al.
Kidney international, 86(1), 199-207 (2014-03-14)
During an outbreak of methanol poisonings in the Czech Republic in 2012, we were able to study methanol and formate elimination half-lives during intermittent hemodialysis (IHD) and continuous veno-venous hemodialysis/hemodiafiltration (CVVHD/HDF) and the relative impact of dialysate and blood flow
Nicolaas A Verwey et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 20(3), 179-187 (2013-07-09)
Abstract Amyloid β-peptide (Aβ) is a key molecule in Alzheimer's disease (AD). Reliable immunohistochemical (IHC) methods to detect Aβ and Aβ-associated factors (AAF) in brain specimens are needed to determine their role in AD pathophysiology. Formic acid (FA) pre-treatment, which
Therese Fagerqvist et al.
Journal of neurochemistry, 126(1), 131-144 (2013-02-01)
Inclusions of intraneuronal alpha-synuclein (α-synuclein) can be detected in brains of patients with Parkinson's disease and dementia with Lewy bodies. The aggregation of α-synuclein is a central feature of the disease pathogenesis. Among the different α-synuclein species, large oligomers/protofibrils have
Sai Duan et al.
Physical chemistry chemical physics : PCCP, 15(13), 4625-4633 (2013-02-21)
Local structures and adsorption energies of a formic acid molecule and its decomposed intermediates (H, O, OH, CO, HCOO, and COOH) on highly electrocatalytically active mushroom-like Au-core@Pd-shell@Pt-cluster nanoparticles with two atomic layers of the Pd shell and stoichiometric Pt coverage
An efficient CoAuPd/C catalyst for hydrogen generation from formic acid at room temperature.
Zhi-Li Wang et al.
Angewandte Chemie (International ed. in English), 52(16), 4406-4409 (2013-03-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service