Skip to Content
MilliporeSigma
All Photos(2)

Documents

F0125

Sigma-Aldrich

N-(4-Amino-2,5-diethoxyphenyl)benzamide

Powder

Synonym(s):

4′-Amino-2′,5′-diethoxybenzanilide, Azoic Diazo No. 20, Fast blue BB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O3
CAS Number:
Molecular Weight:
300.35
Colour Index Number:
37175
Beilstein:
3416889
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

N-(4-Amino-2,5-diethoxyphenyl)benzamide,

form

powder

Quality Level

composition

Dye content, ~95%

mp

98-100 °C

solubility

DMF:H2O (9:1): 10 mg/mL, very slightly hazy, red to red-brown

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCOc1cc(NC(=O)c2ccccc2)c(OCC)cc1N

InChI

1S/C17H20N2O3/c1-3-21-15-11-14(16(22-4-2)10-13(15)18)19-17(20)12-8-6-5-7-9-12/h5-11H,3-4,18H2,1-2H3,(H,19,20)

InChI key

CNXZLZNEIYFZGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service