Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Please contact Customer Service for Availability | $598.00 |
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
certified reference material
agency
ERM®, certified by the BSC
manufacturer/tradename
JRC
application(s)
life science and biopharma
Packaging
set with 5 bars
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Legal Information
ERM is a registered trademark of European Commission
1 of 1
This Item | |||
|---|---|---|---|
| description ERM®, certified reference material, 120 J | description ERM®, certified reference material | description 0.1% Au, 0.5 mm wire, ERM®, certified reference material | description ERM®, certified reference material |
| application(s) life science and biopharma | application(s) - | application(s) - | application(s) environmental |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC |
| agency certified by the BSC, ERM® | agency ERM® | agency ERM® | agency ERM® |
Still not finding the right product?
Explore all of our products under Charpy specimens (impact toughness)
Storage Class
13 - Non Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
