Skip to Content
MilliporeSigma
All Photos(1)

Documents

ERMBB430

Pork fat (OCP′s)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

matrix material

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBB430

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: Hexachlorobenzene, α-HCH, β-HCH, β-HEPO, p,p′-DDT, p,p′-DDD, p,p′-DDE, γ-HCH, Dieldrin, Endrin

Matrix Group: Meat

Legal Information

ERM is a registered trademark of European Commission

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service