Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMAE647

Copper, 63Cu (certified for isotope abundance ratio)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®
ERM®

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

Analysis Note

For more information please see:
ERMAE647

Legal Information

BCR is a registered trademark of European Commission
ERM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service