Skip to Content
MilliporeSigma
All Photos(1)

Documents

ERMAD442K

Lambda DNA solution

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

general analytical

format

single component solution

storage temp.

−20°C

General description

Certified for the DNA mass concentration of LAMBDA DNA in a solution.

ERM-AD442K_cert

ERM-AD442K_report

ERM-AD442K_origin</A

Analysis Note

For more information please see:
ERMAD442K

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service