Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW451300

Sigma-Aldrich

Corning® Gentest UGT Reaction Mix Solution A 2 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

shipped in

dry ice

storage temp.

−20°C

General description

The UGT Reaction Mix comprises two solutions : Solutions A and B. They are sold separately. Solution A includes 25 mM UDPGA, (uridine diphosphate-glucuronic acid) an essential UGT enzyme cofactor. A combination of the two solution serves as a reagent for glucuronidation by microtomes, S9 and recombinant UGT enzymes.
The Corning UGT Reaction Mix is optimized to use with Corning recombinant enzymes and liver microsomes and S9 products. About 200 to 400 enzyme assays can be performed using each system, depending on experimental design.

Legal Information

Corning is a registered trademark of Corning, Inc.
Gentest is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service