Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354242

Sigma-Aldrich

Corning® Cell-Tak Cell Tissue Adhesive (2x5 mg)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

Mytilus edulis (Polyphenolic proteins)

form

liquid (in 5% acetic acid)

mol wt

110-140 kDa

packaging

pkg of 10 mg

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Corning® Cell-Tak cell and tissue adhesive, 10 mg, is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, fluorinated ethylene propylene (FEP) Polymer, and biological materials.

Quality

Dopa: Protein ratio >0.05

Physical form

As a liquid in 5% acetic acid

Other Notes

Biological Source:
Polyphenolic proteins secreted by Mytilus edulis.
Handling:
Recommended concentration - 1-5 μg/cm2 of growth surface depending on cell type (guidelines for coating included with product).

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service