Skip to Content
MilliporeSigma
All Photos(1)

Documents

D5021

Sigma-Aldrich

2,2′-Dihydroxy-1,1′-azonaphthalene-3,3′,6,6′-tetrasulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14N2O14S4
CAS Number:
Molecular Weight:
634.59
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ~75%

SMILES string

Oc1c(cc2cc(ccc2c1N=Nc3c(O)c(cc4cc(ccc34)S(O)(=O)=O)S(O)(=O)=O)S(O)(=O)=O)S(O)(=O)=O

InChI

1S/C20H14N2O14S4/c23-19-15(39(31,32)33)7-9-5-11(37(25,26)27)1-3-13(9)17(19)21-22-18-14-4-2-12(38(28,29)30)6-10(14)8-16(20(18)24)40(34,35)36/h1-8,23-24H,(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36)

InChI key

ZHACZGMWBSRPKU-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service