Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4167

Sigma-Aldrich

4-(p-Dihexadecylaminostyryl)-N-methylpyridinium iodide

Synonym(s):

DiA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C46H79N2I
CAS Number:
Molecular Weight:
787.04
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

storage temp.

−20°C

SMILES string

I.CCCCCCCCCCCCCCCCN(CCCCCCCCCCCCCCCC)c1ccc(\C=C\C2=CC=[N](C)C=C2)cc1

InChI

1S/C46H79N2.HI/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-40-48(41-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)46-36-34-44(35-37-46)32-33-45-38-42-47(3)43-39-45;/h32-39,42-43H,4-31,40-41H2,1-3H3;1H/b33-32+;

InChI key

TXLOTRDAYSHDSG-BQTTXISVSA-N

Application

Fluorescent neuronal tracer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles W Abbott et al.
Cerebral cortex (New York, N.Y. : 1991), 28(8), 2908-2921 (2017-11-07)
Fetal Alcohol Spectrum Disorders, or FASD, represent a range of adverse developmental conditions caused by prenatal ethanol exposure (PrEE) from maternal consumption of alcohol. PrEE induces neurobiological damage in the developing brain leading to cognitive-perceptual and behavioral deficits in the
Tomomi Higashide et al.
Investigative ophthalmology & visual science, 47(7), 2943-2950 (2006-06-27)
To determine whether a scanning laser ophthalmoscope (SLO) is useful for in vivo imaging and counting of rat retinal ganglion cells (RGCs). RGCs of Brown Norway rats were retrogradely labeled bilaterally with the fluorescent dye 4-(4-(dihexadecylamino)styryl)-N-methylpyridinium iodine (DiA). The unilateral
M P Lafuente et al.
Neuroscience, 109(1), 157-168 (2002-01-11)
In adult Sprague-Dawley rats we have investigated retinal ganglion cell survival after transient intervals of retinal ischemia of 30, 45, 60, 90 or 120 min duration, induced by ligature of the ophthalmic vessels. Animals were killed 5, 7, 14, 21
Alessandro E Vercelli et al.
The European journal of neuroscience, 20(2), 495-502 (2004-07-06)
The apical dendrites of the pyramidal neurons of the cerebral cortex form radial bundles in all species and areas. Using microtubule-associated protein (MAP)2 immunostaining and Voronoi tessellation analysis in the rat visual cortex, we obtained objective criteria to define dendritic
Mark W Doyle et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(18), 8222-8229 (2002-09-12)
Although the central terminals of cranial visceral afferents express vanilloid receptor 1 (VR1), little is known about their functional properties at this first synapse within the nucleus tractus solitarius (NTS). Here, we examined whether VR1 modulates afferent synaptic transmission. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service