Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2385

Sigma-Aldrich

Diphenylamine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
Beilstein:
508755
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.82 (vs air)

vapor pressure

1 mmHg ( 108 °C)

Assay

≥98%

autoignition temp.

1175 °F

bp

302 °C (lit.)

mp

50-53 °C (lit.)

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quality

Purified

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter J Jessen et al.
The Journal of clinical investigation, 123(1), 340-347 (2012-12-12)
Neurofibromatosis type 1 (NF1) patients develop benign neurofibromas and malignant peripheral nerve sheath tumors (MPNST). These incurable peripheral nerve tumors result from loss of NF1 tumor suppressor gene function, causing hyperactive Ras signaling. Activated Ras controls numerous downstream effectors, but
Sarah G Pati et al.
Environmental science & technology, 46(21), 11844-11853 (2012-09-29)
Dioxygenation of aromatic rings is frequently the initial step of biodegradation of organic subsurface pollutants. This process can be tracked by compound-specific isotope analysis to assess the extent of contaminant transformation, but the corresponding isotope effects, especially for dioxygenation of
M Francklin Philips et al.
Journal of hazardous materials, 237-238, 46-54 (2012-09-12)
Poly(diphenylamine-co-2-aminobenzonitrile) (P(DPA-co-2ABN)), a cyano group containing conducting polyaniline derivative, has been electrodeposited developed as the new material and utilized for the simultaneous electrochemical determination of trace levels of cadmium (Cd(2+)) and lead (Pb(2+)). P(DPA-co-2ABN) film preconcentrates effectively through cyano chelation
Margot Van der Jeught et al.
Stem cells and development, 22(2), 296-306 (2012-07-13)
In embryonic stem cell culture, small molecules can be used to alter key signaling pathways to promote self-renewal and inhibit differentiation. In mice, small-molecule inhibition of both the FGF/MEK/Erk and the GSK3β pathways during preimplantation development suppresses hypoblast formation, and
Tiffany Chang et al.
The Journal of clinical investigation, 123(1), 335-339 (2012-12-12)
Children with neurofibromatosis type 1 (NF1) are predisposed to juvenile myelomonocytic leukemia (JMML), an aggressive myeloproliferative neoplasm (MPN) that is refractory to conventional chemotherapy. Conditional inactivation of the Nf1 tumor suppressor in hematopoietic cells of mice causes a progressive MPN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service