Skip to Content
MilliporeSigma
All Photos(1)

Documents

D1945000

Dihydroergocristine mesilate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dihydroergocristine methanesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H41N5O5 · CH4O3S
CAS Number:
Molecular Weight:
707.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dihydroergocristine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.[H][C@@]12Cc3c[nH]c4cccc(c34)[C@@]1([H])C[C@H](CN2C)C(=O)N[C@@]5(O[C@]6(O)N([C@@H](Cc7ccccc7)C(=O)N8CCC[C@@]68[H])C5=O)C(C)C

InChI

1S/C35H41N5O5.CH4O3S/c1-20(2)34(37-31(41)23-16-25-24-11-7-12-26-30(24)22(18-36-26)17-27(25)38(3)19-23)33(43)40-28(15-21-9-5-4-6-10-21)32(42)39-14-8-13-29(39)35(40,44)45-34;1-5(2,3)4/h4-7,9-12,18,20,23,25,27-29,36,44H,8,13-17,19H2,1-3H3,(H,37,41);1H3,(H,2,3,4)/t23-,25-,27-,28+,29+,34-,35+;/m1./s1

InChI key

SPXACGZWWVIDGR-SPZWACKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dihydroergocristine mesilate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woong Mo Kim et al.
European journal of pharmacology, 793, 112-118 (2016-11-14)
Controlling extracellular glutamate level in a physiological range is important to maintain normal sensory transmission. Here, we investigated the paradoxical action of glutamate transporters in the rat formalin test to elucidate a possible role of inversely operating transporters in its
Hyung Gon Lee et al.
Neuroscience letters, 583, 103-107 (2014-09-23)
The purpose of this study was to validate the effects of tianeptine on serotonergic and noradrenergic neurotransmission in a rat model of neuropathic pain. Neuropathic pain was induced by ligating the L5 and L6 spinal nerves in male Sprague-Dawley rats
Hye Jin Jeong et al.
The Korean journal of pain, 24(4), 179-184 (2012-01-06)
The analgesic mechanisms of cyclooxygenase (COX)-2 inhibitors have been explained mainly on the basis of the inhibition of prostaglandin biosynthesis. However, several lines of evidence suggest that their analgesic effects are mediated through serotonergic or adrenergic transmissions. We investigated the
Julio Alonso-Padilla et al.
PLoS neglected tropical diseases, 9(1), e0003493-e0003493 (2015-01-24)
Chagas disease, caused by the protozoan parasite Trypanosoma cruzi, represents a very important public health problem in Latin America where it is endemic. Although mostly asymptomatic at its initial stage, after the disease becomes chronic, about a third of the
Wentao Lyu et al.
Frontiers in cellular and infection microbiology, 8, 191-191 (2018-06-27)
A rise in antimicrobial resistance demands novel alternatives to antimicrobials for disease control and prevention. As an important component of innate immunity, host defense peptides (HDPs) are capable of killing a broad spectrum of pathogens and modulating a range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service