Skip to Content
MilliporeSigma
All Photos(1)

Documents

D1300000

Diethylcarbamazine citrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Diethylcarbamazine citrate salt, N,N-Diethyl-4-methyl-1-piperazinecarboxamide citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H21N3O · C6H8O7
CAS Number:
Molecular Weight:
391.42
Beilstein:
3836352
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diethylcarbamazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1

InChI

1S/C10H21N3O.C6H8O7/c1-4-12(5-2)10(14)13-8-6-11(3)7-9-13;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-9H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

PGNKBEARDDELNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethylcarbamazine citrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfram Gottfried Metzger et al.
The Lancet. Infectious diseases, 14(4), 353-357 (2013-12-18)
More than 10 million people in western and central Africa are estimated to be infected with Loa loa filarial nematodes. Like most other infectious diseases, L loa filariasis (loiasis) covers a wide range of symptoms. Severe complications have been reported;
M García-Hernández et al.
International immunopharmacology, 23(1), 113-120 (2014-08-26)
We tested whether diethylcarbamazine (DEC) or ivermectin (IVM), both antiparasitic drugs with reported immunomodulatory properties, were able to affect the immune system to potentiate host defense mechanisms and protect against actinomycetoma in a mouse model. Male BALB/c mice of 10-12
Mark S Schmidt et al.
Journal of pharmaceutical and biomedical analysis, 98, 307-310 (2014-07-01)
A sensitive and selective liquid chromatographic method using mass spectrometric detection was developed for the determination of diethylcarbamazine (DEC) in human plasma. DEC and its stable isotope internal standard d3-DEC were extracted from 0.25mL of human plasma using solid phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service