CRMUFRKR
Freezing Point Standard
ASTM D 2386 (-54.1 °C)
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
according to ASTM® 2386
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
mp
-49 °C
application(s)
jet fuel
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This product is a certified reference material (CRM), manufactured under UKAS accreditation
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
84.2 - 158.0 °F - closed cup
Flash Point(C)
29 - 70 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service