Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM170

Supelco

PAHs - Clay Soil

pkg of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM170-100

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

PAHs Clay Soil has been used as a certified soil reference material for the quantification of polycyclic aromatic hydrocarbons in soil samples using gas chromatography (GC) technique.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Benzo[a]pyrene
    Benzo[b]fluoranthene
    Benzo[ghi]perylene
    Benzo[k]fluoranthene
    Chrysene
    Fluoranthene
    Fluorene
    Naphthalene
    Phenanthrene
    Pyrene
    See All (13)

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biljana Milenkovic et al.
Chemosphere, 245, 125610-125610 (2019-12-22)
After NATO bombing of Serbia in 1999, UNEP has identified Kragujevac as one of the four heavily polluted environmental "hot spots". Damaging of industrial and military targets caused the release of substantial amounts of hazardous chemical substances into the environment.
Rapid extraction method of polycyclic aromatic compounds in soil using basic silica selective pressurized liquid extraction
Titaley AI, et al.
Journal of Chromatography A (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service