Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CCW5078

Millipore

SGQ + Broth in flasks

Liquid medium in Bottle for low acid beverage spoiling microorganisms, specially Dekkera spp., suitable for microbiology

Synonym(s):

SGQB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

sterility

sterile

form

liquid

shelf life

12 mo. at 8‑25 °C

feature

ready-to-use

packaging

box of 10 bottles

storage condition

(Tightly closed)

parameter

450 mL filling volume

bottle capacity

500 mL

capacity

2 mL

color

pale yellow

pH

4.3±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Brettanomyces/Dekkera spp.

General description

The SGQ + Broth allows the growth of organisms that can spoil low-acid beverages, especially Dekkera spp.

Application

SGQ + Broth in flasks are used to detect low acid beverage spoiling microorganisms, specially Dekkera spp.

Components

  • 1 box with 10 bottles of 500 mL
  • Injectable cap: Plastic screw inner cap

Other Notes

  • Using syringe needles with a diameter greater than 0.8 mm is not recommended.
  • Liquid medium formulated according to the specifications of the customer.
  • Use the medium according to the intended purpose, samples, and validated methods.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service