Skip to Content
MilliporeSigma
All Photos(1)

Documents

C2857

Sigma-Aldrich

Cupric Sulfate

meets USP testing specifications

Synonym(s):

Copper(II) sulfate pentahydrate, Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
meets USP testing specifications

vapor pressure

7.3 mmHg ( 25 °C)

form

crystals

mp

110 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nelson Durán et al.
SpringerPlus, 3, 645-645 (2014-12-09)
In the present study, semi-purified laccase from Trametes versicolor was applied for the synthesis of silver nanoparticles, and the properties of the produced nanoparticles were characterized. All of the analyses of the spectra indicated silver nanoparticle formation. A complete characterization
William T Triplett et al.
Magnetic resonance in medicine, 72(1), 8-19 (2013-09-06)
The relationship between fat fractions (FFs) determined based on multiple TE, unipolar gradient echo images and (1) H magnetic resonance spectroscopy (MRS) was evaluated using different models for fat-water decomposition, signal-to-noise ratios, and excitation flip angles. A combination of single-voxel
David Rehnlund et al.
Nanoscale, 7(32), 13591-13604 (2015-07-25)
Electrodes composed of freestanding nano- and microrods composed of stacked layers of copper and cuprous oxide have been fabricated using a straightforward one-step template-assisted pulsed galvanostatic electrodeposition approach. The approach provided precise control of the thickness of each individual layer
Pieter Spincemaille et al.
Toxicology and applied pharmacology, 280(2), 345-351 (2014-08-20)
Wilson disease (WD) is caused by accumulation of excess copper (Cu) due to a mutation in the gene encoding the liver Cu transporter ATP7B, and is characterized by acute liver failure or cirrhosis and neuronal cell death. We investigated the
A S Singha et al.
International journal of biological macromolecules, 67, 409-417 (2014-04-08)
Use of biological macromolecules for wastewater remediation process has become the topic of intense research mostly driven by growing concerns about the depletion of petroleum oil reserves and environmental problems. So in view of technological significance of cellulosic biopolymers in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service