Skip to Content
MilliporeSigma
All Photos(1)

Documents

C1176

Millipore

Corn meal agar

microbiology tested, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

suitability

microbiology tested
Brucella spp.
Candida spp.
molds
yeasts

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

Application

Recommended for enrichment, isolation and cultivation of Brucella species.

Preparation Note

Solve 17.0 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service