Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP639

Form-2′,4′-xylidide

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

N-(2,4-Dimethylphenyl)formamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$346.00

$346.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$346.00

About This Item

Linear Formula:
(CH3)2C6H3NHCHO
CAS Number:
Molecular Weight:
149.19
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$346.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

amitraz

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

114-118 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

−20°C

SMILES string

[H]C(=O)Nc1ccc(C)cc1C

InChI

1S/C9H11NO/c1-7-3-4-9(10-6-11)8(2)5-7/h3-6H,1-2H3,(H,10,11)

InChI key

JOFDPSBOUCXJCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Form-2′,4′-xylidide BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service