Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP583

Meptazinol hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

3-(3-ethylhexahydro-1-methyl-1H-azepin-3-yl)-phenol hydrochloride, IL-22811 hydrochloride, WY-22811 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$310.00

$310.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$310.00

About This Item

Empirical Formula (Hill Notation):
C15H23NO· HCl
CAS Number:
Molecular Weight:
269.81
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$310.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

meptazinol

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCC1(CCCCN(C)C1)c2cccc(O)c2

InChI

1S/C15H23NO.ClH/c1-3-15(9-4-5-10-16(2)12-15)13-7-6-8-14(17)11-13;/h6-8,11,17H,3-5,9-10,12H2,1-2H3;1H

InChI key

MPJUSISYVXABBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meptazinol hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Meptazinol Injection
  • Meptazinol Tablets
  • Biochem/physiol Actions

    Meptazinol entered the human pharmaceutical market as a racemic mixture in the 1980s for use as an analgesic.
    Meptazinol entered the human pharmaceutical market as a racemic mixture in the 1980s for use as an analgesic. Its pharmacology is not completely understood; however, its analgesic properties are mostly due to its partial agonism at the mu1 opioid receptor. Due to its partial agonism, Meptazinol antagonizes morphine dependence in vivo. Its advantage over other opiates is its reduced capacity to cause addition and respiratory depression, also due to its intrinsic activity as a partial agonist. Meptazinol has been found to have additional activity as an acetylcholinesterase (AChE) inhibitor, particularly in its (-) enantiomeric form, which may partially explain its analgesic properties. AChE inhibitors are used to treat Alzheimer′s disease, providing additional interest in this compound.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service