Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP001

Acepromazine maleate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Acetopromazine maleate salt, 2-Acetyl-10-(3-dimethylaminopropyl)phenothiazine maleate salt, Acepromazine maleate salt

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$310.00

$310.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$310.00

About This Item

Empirical Formula (Hill Notation):
C19H22N2OS · C4H4O4
CAS Number:
Molecular Weight:
442.53
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$310.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

acepromazine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(C)=O

InChI

1S/C19H22N2OS.C4H4O4/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)23-19;5-3(6)1-2-4(7)8/h4-5,7-10,13H,6,11-12H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

FQRHOOHLUYHMGG-BTJKTKAUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support

Application

Acepromazine maleate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Acepromazine Injection
  • Acepromazine Tablets
  • Acepromazine Maleate
  • Biochem/physiol Actions

    Acepromazine is a phenothiazine antipsychotic comonly used as a veterinary drug (horses, dogs and cats). The compound is an analogue antipsychotic drug chlorpromazine (#C8138). The drug is thought to block receptors of dopamine in the brain. Recently it was discover that the compound inhibits ABCG2 transported protein, which overexpression in tumors is associated with drug resistance.
    Atypical neuroleptic.

    Packaging

    Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    related product

    Pictograms

    Skull and crossbones

    Signal Word

    Danger

    Hazard Statements

    Precautionary Statements

    Hazard Classifications

    Acute Tox. 3 Oral - STOT SE 3

    Target Organs

    Central nervous system

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service