Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR660

Ethanol in water (SNIF)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

450 ML
$1,520.00

$1,520.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
450 ML
$1,520.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$1,520.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

isotopic purity

 2H

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Analysis Note

For more information please see:
BCR660

Legal Information

BCR is a registered trademark of European Commission

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service