Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR342

Benzo[a]fluorenone

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$333.00

$333.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$333.00

About This Item

Empirical Formula (Hill Notation):
C17H10O
CAS Number:
Molecular Weight:
230.26
Beilstein:
1960212
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$333.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

O=C1c2ccccc2-c3ccc4ccccc4c13

InChI

1S/C17H10O/c18-17-15-8-4-3-7-13(15)14-10-9-11-5-1-2-6-12(11)16(14)17/h1-10H

InChI key

RNICURKFVSAHLQ-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR342

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service