Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

B8800

Sigma-Aldrich

Bacitracin zinc salt

from Bacillus licheniformis, meets USP testing specifications

Synonym(s):

Zinc bacitracin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H101N17O16SZn
CAS Number:
Molecular Weight:
1486.07
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.76

biological source

Bacillus licheniformis

Quality Level

Agency

USP/NF
meets USP testing specifications

form

powder

mp

250 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Zn++].CCC(C)C(N)C1=NCC(S1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@H](CCC([O-])=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@H]2CCCCNC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](CC([O-])=O)NC(=O)[C@H](Cc3cnc[nH]3)NC(=O)[C@@H](Cc4ccccc4)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC2=O)[C@@H](C)CC

InChI

1S/C66H103N17O16S.Zn/c1-9-35(6)52(69)66-72-32-48(100-66)63(97)80-43(26-34(4)5)59(93)75-42(22-23-50(85)86)58(92)83-53(36(7)10-2)64(98)76-40-20-15-16-25-71-55(89)46(29-49(68)84)78-62(96)47(30-51(87)88)79-61(95)45(28-39-31-70-33-73-39)77-60(94)44(27-38-18-13-12-14-19-38)81-65(99)54(37(8)11-3)82-57(91)41(21-17-24-67)74-56(40)90;/h12-14,18-19,31,33-37,40-48,52-54H,9-11,15-17,20-30,32,67,69H2,1-8H3,(H2,68,84)(H,70,73)(H,71,89)(H,74,90)(H,75,93)(H,76,98)(H,77,94)(H,78,96)(H,79,95)(H,80,97)(H,81,99)(H,82,91)(H,83,92)(H,85,86)(H,87,88);/q;+2/p-2/t35?,36-,37-,40-,41+,42+,43-,44+,45-,46-,47+,48?,52?,53-,54-;/m0./s1

InChI key

NKULICGUDKGGRL-FCHFGNCGSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Bacitracin is used to study the disruption of bacterial cell wall synthesis at the level of peptidoglycan biosynthesis and isoprenyl metabolism. Used to study the biosynthesis of sterols and squalene.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Wolf et al.
Antimicrobial agents and chemotherapy, 56(11), 5907-5915 (2012-09-12)
L-forms are cell wall-deficient bacteria that can grow and proliferate in osmotically stabilizing media. Recently, a strain of the Gram-positive model bacterium Bacillus subtilis was constructed that allowed controlled switching between rod-shaped wild-type cells and corresponding L-forms. Both states can
Alina Olender et al.
Medycyna doswiadczalna i mikrobiologia, 64(1), 1-10 (2012-07-20)
The genus Streptococcus comprises a number of species characterized by a differential pathogenic potential. These bacteria can be considered as members of microbial physiological flora but they can also cause mild infections or severe, life threatening conditions. The majority of
Vanina Dengler et al.
FEMS microbiology letters, 333(2), 109-120 (2012-05-30)
The Staphylococcus aureus cell wall stress stimulon (CWSS) is activated by cell envelope-targeting antibiotics or depletion of essential cell wall biosynthesis enzymes. The functionally uncharacterized S. aureus LytR-CpsA-Psr (LCP) proteins, MsrR, SA0908 and SA2103, all belong to the CWSS. Although not
Shanna Spring et al.
Dermatitis : contact, atopic, occupational, drug, 23(5), 210-213 (2012-09-27)
Topical medicaments are a common cause of allergic contact dermatitis. This study will evaluate the prevalence of contact allergy to a wide array of topical medicaments at the Ottawa Patch Test Clinic. The objectives of this study are to report
Audrey Charlebois et al.
PloS one, 7(9), e44449-e44449 (2012-09-13)
Phenotypic bacitracin resistance has been reported in Clostridium perfringens. However, the genes responsible for the resistance have not yet been characterized. Ninety-nine C. perfringens isolates recovered from broilers and turkeys were tested for phenotypic bacitracin resistance. Bacitracin MIC(90) (>256 µg/ml)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service