Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ARK2219

SAFC

PEP TCHA

Pharma Manufacturing

Synonym(s):

Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt, 2-(Phosphonooxy)-2-propenoic acid tri(cyclohexylammonium) salt, PEP-3CHA, Phosphoenolpyruvic acid tris(cyclohexylamine) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5O6P · 3C6H13N
CAS Number:
Molecular Weight:
465.56
Beilstein:
3752336
MDL number:
UNSPSC Code:
12352106

Quality Level

storage temp.

-10 to -25°C

SMILES string

NC1CCCCC1.NC2CCCCC2.NC3CCCCC3.OC(=O)C(=C)OP(O)(O)=O

InChI

1S/3C6H13N.C3H5O6P/c3*7-6-4-2-1-3-5-6;1-2(3(4)5)9-10(6,7)8/h3*6H,1-5,7H2;1H2,(H,4,5)(H2,6,7,8)

InChI key

MJKYGUXBFYGLLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phospho(enol)pyruvic acid (PEP) is involved in glycolysis and gluconeogenesis. In glycolysis, PEP is metabolized by pyruvate kinase to yield pyruvate. In plants, PEP is involved in the formation of aromatic amino acids as well as in the carbon fixation pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service