Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6404

Sigma-Aldrich

Acetic anhydride

reagent grade, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
Beilstein:
385737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

3.5 (vs air)

vapor pressure

10 mmHg ( 36 °C)
4 mmHg ( 20 °C)

Assay

≥98%

autoignition temp.

629 °F

expl. lim.

10.3 %

refractive index

n20/D 1.390 (lit.)

bp

138-140 °C (lit.)

mp

−73 °C (lit.)

solubility

H2O: slightly soluble

density

1.08 g/mL (lit.)

SMILES string

CC(=O)OC(C)=O

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnese Po et al.
Stem cells international, 2017, 5274171-5274171 (2017-03-17)
Cell development is regulated by a complex network of mRNA-encoded proteins and microRNAs, all funnelling onto the modulation of self-renewal or differentiation genes. How intragenic microRNAs and their host genes are transcriptionally coregulated and their functional relationships for the control
Evelina Miele et al.
Molecular oncology, 15(2), 523-542 (2020-09-14)
Persistent mortality rates of medulloblastoma (MB) and severe side effects of the current therapies require the definition of the molecular mechanisms that contribute to tumor progression. Using cultured MB cancer stem cells and xenograft tumors generated in mice, we show
Lakshmi Pillai-Kastoori et al.
PLoS genetics, 10(7), e1004491-e1004491 (2014-07-11)
Ocular coloboma is a sight-threatening malformation caused by failure of the choroid fissure to close during morphogenesis of the eye, and is frequently associated with additional anomalies, including microphthalmia and cataracts. Although Hedgehog signaling is known to play a critical
David G Simmons et al.
BMC genomics, 9, 352-352 (2008-07-30)
The Prolactin (PRL) hormone gene family shows considerable variation among placental mammals. Whereas there is a single PRL gene in humans that is expressed by the pituitary, there are an additional 22 genes in mice including the placental lactogens (PL)
Heidi I Chen et al.
Development (Cambridge, England), 141(23), 4500-4512 (2014-11-08)
Identifying coronary artery progenitors and their developmental pathways could inspire novel regenerative treatments for heart disease. Multiple sources of coronary vessels have been proposed, including the sinus venosus (SV), endocardium and proepicardium, but their relative contributions to the coronary circulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service