Skip to Content
MilliporeSigma
All Photos(1)

Documents

A3766

Supelco

Antazoline phosphate salt

analytical standard, for drug analysis

Synonym(s):

2-(N-Benzylanilinomethyl)-2-imidazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H19N3 · H3PO4
CAS Number:
Molecular Weight:
363.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

OP(O)(O)=O.C1CN=C(CN(Cc2ccccc2)c3ccccc3)N1

InChI

1S/C17H19N3.H3O4P/c1-3-7-15(8-4-1)13-20(14-17-18-11-12-19-17)16-9-5-2-6-10-16;1-5(2,3)4/h1-10H,11-14H2,(H,18,19);(H3,1,2,3,4)

InChI key

DUIGUKRYYAGJAF-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Imidazoline agonist; more potent than efaroxan in inducing insulin release from β cells; H1 histamine receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Berdeu et al.
European journal of pharmacology, 324(2-3), 233-239 (1997-04-18)
In vivo effects of an imidazoline devoid of alpha2-adrenoceptor antagonistic properties, antazoline, on insulin secretion and glycemia were investigated both in fasted rats and dogs. In both species, antazoline (1.5 mg/kg i.v.) transiently increased insulinemia without affecting basal plasma glucose
D Berdeu et al.
European journal of pharmacology, 275(1), 91-98 (1995-02-24)
The relative potencies of imidazoline compounds to induce insulin secretion and vascular resistance were compared in the isolated perfused rat pancreas. On insulin secretion, only the two imidazolines, antazoline and efaroxan, induced a concentration-dependent response, antazoline being 10 times more
R Wertheimer et al.
Klinische Monatsblatter fur Augenheilkunde, 210(2), 93-96 (1997-02-01)
Allergic conjunctivitis is one of the most frequent allergic diseases of the anterior eye segment. This multicentre, clinical trial was an investigation to compare the antiallergic efficacy, local tolerance and safety of Antazolin/Tetryzolin eye drops and Levocabastine eye drops. 69
D Berdeu et al.
European journal of pharmacology, 254(1-2), 119-125 (1994-03-11)
The effects of imidazolines and derivatives were studied on insulin secretion and vascular resistance in the isolated perfused rat pancreas. On insulin secretion, two imidazoline alpha 2-adrenoceptor antagonists, efaroxan (1-100 microM) and RX821002 (10 microM), had a stimulating response; however
Mariusz Swiader et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 14(4), 307-318 (2004-05-28)
Experimental studies have indicated that the central histaminergic system plays an important role in the inhibition of seizures through the stimulation of histamine H1 receptors. H1 receptor antagonists, including classical antiallergic drugs, occasionally may induce convulsions in healthy children and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service