Skip to Content
MilliporeSigma
All Photos(2)

Documents

A3174

Sigma-Aldrich

Bovine Serum Albumin solution

30% in saline, high avidity, contains azide and caprylate, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

28-32% (biuret)

form

solution

quality

high avidity, contains azide and caprylate

purified by

heat shock fractionation

contains

0.1% sodium azide as preservative
10-15 mg/gram protein octanoic acid as stabilizer

origin

USA origin

concentration

28-32% (biuret)
30% in saline

technique(s)

blocking: suitable

UniProt accession no.

foreign activity

BT Virus, none detected
VSV Virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin (BSA) belongs to the serum albumin family. It comprises three domains with two sub-domains under each. It is an α-helical, globular and non-glycosylated protein. BSA has 17-disulfide bonds.

Application

Bovine Serum Albumin solution has been used in the blocking of nonspecific binding in brain sections prior to light microscopy studies.

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is a transporter for drugs, hormones and fatty acids. It is a crucial component of the cell culture media and favors embryonic stem cells (hESC) differentiation. BSA displays high solubility and is exploited in the cosmetic and pharmaceutical industry.It is used as a blocking agent in enzyme-linked immunosorbent assay (ELISA).

Analysis Note

Tested for avidity with known incomplete antibody (not saline agglutinable).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cystic fibrosis transmembrane conductance regulator protein expression in the male excretory duct system during development
Marcorelles P, et al.
Human Pathology, 43(3), 390-397 (2012)
Binding of fatty acid amide amphiphiles to bovine serum albumin: role of amide hydrogen bonding
Ghosh S and Dey J
The Journal of Physical Chemistry B, 119(25), 7804-7815 (2015)
Enzyme-linked immunosorbent assay (ELISA) and blocking with bovine serum albumin (BSA)?not all BSAs are alike
Xiao Y and Isaacs SN
Journal of Immunological Methods, 384(1-2), 148-151 (2012)
Albumin and mammalian cell culture: implications for biotechnology applications
Francis GL
Cytotechnology, 62(1), 1-16 (2010)
Structural changes during the unfolding of Bovine serum albumin in the presence of urea: A small-angle neutron scattering study
Das A, et al.
Pramana, 63(2), 363-368 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service