Skip to Content
MilliporeSigma
All Photos(1)

Documents

A1225000

Alfadex

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

α-Cyclodextrin, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
Beilstein:
79627
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

description

API family: alfadex

API family

alfadex, cyclodextrin

manufacturer/tradename

EDQM

mp

>278 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-/m1/s1

InChI key

HFHDHCJBZVLPGP-RWMJIURBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alfadex EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidetoshi Arima
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 124(7), 451-464 (2004-07-06)
Due to the growing concerns over the toxicity and immunogenicity of viral DNA delivery systems, DNA delivery via nonviral routes has become more desirable and advantageous. In particular, polycation complexes with DNA (polyplex) are attractive nonviral vectors. To design novel
Martha Mullally et al.
Canadian journal of physiology and pharmacology, 92(9), 758-764 (2014-08-21)
The mode of action of the anxiolytic medicinal plant Souroubea sympetala was investigated to test the hypothesis that extracts and the active principle act at the pharmacologically important GABAA-benzodiazepine (GABAA-BZD) receptor. Leaf extracts prepared by ethyl acetate extraction or supercritical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service