Skip to Content
MilliporeSigma
All Photos(1)

Documents

A0593

Sigma-Aldrich

Alprostadil

meets USP testing specifications

Synonym(s):

Prostaglandin E1, (11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
Beilstein:
5294062
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

meets USP testing specifications

Assay

95.0-105.0% dry basis

form

powder

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1

InChI key

GMVPRGQOIOIIMI-DWKJAMRDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Costa et al.
Drugs, 72(17), 2243-2254 (2012-11-23)
The global burden of erectile dysfunction (ED) is increasing. It is estimated that 8-19% of men in Europe have ED and that by 2025 the prevalence of ED worldwide will reach 322 million. The gold standard therapy for ED is
Adalet Demir et al.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery, 47(6), 974-983 (2014-09-05)
Donor to recipient (D/R) matching in lung transplantation (LTx) is usually directed by blood group (identity or compatibility) and predicted total lung capacity (pTLC) based on height and age. Other donor (D) and recipient (R) characteristics such as cytomegalovirus (CMV)
Kristine J Guleserian et al.
The Journal of thoracic and cardiovascular surgery, 145(1), 206-213 (2012-12-19)
Bilateral pulmonary artery banding with or without ductal stenting has been performed as a resuscitative intervention for patients considered at too high risk for conventional single ventricle palliation. The purpose of the present study was to determine the outcomes using
Miriam Furne et al.
Fish & shellfish immunology, 34(3), 770-777 (2013-01-08)
Primary head kidney leukocytes from Atlantic cod were isolated to evaluate the use of arachidonic acid and eicosapentaenoic acid by cyclooxygenases and the production of prostaglandins E₂ and E₃. The expression of cyclooxygenase genes and selected interleukin genes like Interleukin
Lisbeth Svensson et al.
Infection and immunity, 82(10), 4307-4314 (2014-07-30)
Platelet activation and aggregation have been reported to occur in response to a number of Gram-positive pathogens. Here, we show that platelet aggregates induced by Streptococcus pyogenes were unstable and that viable bacteria escaped from the aggregates over time. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service