Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96602

Sigma-Aldrich

Bis(cyclopentadienyl)zirconium bis(trifluoromethanesulfonate) tetrahydrofuran complex

purum, ≥97.0%

Synonym(s):

Di(cyclopentadienyl)zirconium bis(trifluoromethanesulfonate) THF complex, Zirconocene bis(trifluoromethanesulfonate) THF complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F6O6S2Zr · C4H8O
CAS Number:
Molecular Weight:
591.65
MDL number:
UNSPSC Code:
12352300

grade

purum

Assay

≥97.0%

SMILES string

[Zr++].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.C3CCOC3.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Other Notes

Catalyst for Diels-Alder reactions; Catalyst for Mukaiyama cross-aldol reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.K. Hollis et al.
Tetrahedron Letters, 33, 6423-6423 (1992)
T.K. Hollis et al.
Organometallics, 11, 2745-2745 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service