Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95851

Supelco

1,3,5-Tri-tert-butyl-2-nitrosobenzene

for ESR-spectroscopy, >98.0% (HPLC)

Synonym(s):

2,4,6-Tri-tert-butylnitrosobenzene, 2-Nitroso-1,3,5-tri-tert-butylbenzene, TTBNB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29NO
CAS Number:
Molecular Weight:
275.43
Beilstein:
1880006
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for ESR-spectroscopy

Assay

>98.0% (HPLC)

form

solid

technique(s)

UV/Vis spectroscopy: suitable

mp

168-173 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(c(N=O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C18H29NO/c1-16(2,3)12-10-13(17(4,5)6)15(19-20)14(11-12)18(7,8)9/h10-11H,1-9H3

InChI key

OSICDPWAPKXXHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Spin trapping reagent for ESR spectrometry; the cation radical is prepared by electrolysis in MeNO2/MeCN

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Cauquis et al.
J. Chem. Phys. , 47, 4642-4642 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service