1 U corresponds to the amount of enzyme which liberates 1 μmol remazol brilliant blue R at pH 6.0 and 40 °C (remazol brilliant blue R-xylan, Cat. No. 66960, as substrate)
Biotechnology and bioengineering, 110(11), 2959-2969 (2013-05-02)
Corynebacterium glutamicum is an important industrial organism that is widely used in the production of amino acids, nucleotides and vitamins. To extend its product spectrum and improve productivity, C. glutamicum needs to undergo further engineering, including the development of applicable
We propose a new process of unified aerobic delignification and anaerobic saccharification and fermentation of wood by a single microorganism, the white-rot fungus Phlebia sp. MG-60. This fungus is able to selectively degrade lignin under aerobic solid state fermentation conditions
Journal of agricultural and food chemistry, 60(51), 12516-12524 (2012-11-09)
A novel gene fragment containing a xylanase was identified from a Holstein cattle rumen metagenomic library. The novel xylanase (Xyln-SH1) belonged to the glycoside hydrolase family 10 (GH10) and exhibited a maximum of 44% identity to the glycoside hydrolase from
A novel xylanase gene, xyn10A, was cloned from Flavobacterium johsoniae, overexpressed in a flavobacterial expression system, the recombinant enzyme purified by Ni-affinity chromatography, and enzyme structure and activity analyzed. Xyn10A was found to be a modular xylanase with an Fn3